- Conjugates of cytochrome
Conjugates of cytochrome
Experimental and therapeutic medicine (2013-10-19)
Patrick Imesch, David Scheiner, Emese Szabo, Daniel Fink, André Fedier
PMID24137266
ABSTRACT
Polycationic cell-penetrating peptides (CPPs) deliver macromolecules into cells without losing the functional properties of the cargoed macromolecule. The aim of this study was to determine whether exogenous cytochrome