Merck
Hydrogels for biomedical applications.
Advanced drug delivery reviews (2002-01-05)
Allan S Hoffman
ABSTRACT
This article reviews the composition and synthesis of hydrogels, the character of their absorbed water, and permeation of solutes within their swollen matrices. The most important properties of hydrogels relevant to their biomedical applications are also identified, especially for use of hydrogels as drug and cell carriers, and as tissue engineering matrices.