Merck
  • Vibrational spectra and assignments of 2-amino-5-iodopyridine by ab initio Hartree-Fock and density functional methods.
Vibrational spectra and assignments of 2-amino-5-iodopyridine by ab initio Hartree-Fock and density functional methods.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-10-05)
N Sundaraganesan, C Meganathan, B Anand, B Dominic Joshua, Christine Lapouge
ABSTRACT
The Fourier transform Raman and Fourier transform infrared spectra of 2-amino-5-iodopyridine were recorded in the solid phase. The equilibrium geometry, harmonic vibrational frequencies, infrared intensities and Raman scattering activities were calculated by HF and DFT (B3LYP) methods with the 6-31G(d,p) basis set for C, N, H and LANL2DZ pseudopotential for I. The scaled theoretical wavenumbers showed very good agreement with the experimental ones. A detailed interpretation of the infrared and Raman spectra of 2-amino-5-iodopyridine is reported on the basis of the calculated potential energy distribution. The theoretical spectrograms for the IR spectrum of the title molecule have been constructed.
Sigma-Aldrich
2-Amino-5-iodopyridine, 98%