- Identification and characterization of the gene encoding the human phosphopantetheine adenylyltransferase and dephospho-CoA kinase bifunctional enzyme (CoA synthase).
Identification and characterization of the gene encoding the human phosphopantetheine adenylyltransferase and dephospho-CoA kinase bifunctional enzyme (CoA synthase).
The Biochemical journal (2002-05-08)
Suren Aghajanian, D Margaret Worrall
PMID11994049
ABSTRACT
The final two enzymes in the CoA biosynthetic pathway, phosphopantetheine adenylyltransferase (PPAT; EC 2.7.7.3) and dephospho-CoA kinase (DPCK; EC 2.7.1.24), are separate proteins in prokaryotes, but exist as a bifunctional enzyme in pig liver. In the present study we have obtained sequence information from purified pig-liver enzyme, and identified the corresponding cDNA in a number of species. The human gene localizes to chromosome 17q12-21 and contains regions with sequence similarity to the monofunctional Escherichia coli DPCK and PPAT. The recombinant 564-amino-acid human protein confirmed the associated transferase and kinase activities, and gave similar kinetic properties to the wild-type pig enzyme.