Merck
  • Quantitative analysis of cyclic dimer fatty acid content in the dimerization product by proton NMR spectroscopy.
Quantitative analysis of cyclic dimer fatty acid content in the dimerization product by proton NMR spectroscopy.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-05-15)
Kyun Joo Park, Minyoung Kim, Seunghwan Seok, Young-Wun Kim, Do Hyun Kim
ABSTRACT
In this work, (1)H NMR is utilized for the quantitative analysis of a specific cyclic dimer fatty acid in a dimer acid mixture using the pseudo-standard material of mesitylene on the basis of its structural similarity. Mesitylene and cyclic dimer acid levels were determined using the signal of the proton on the cyclic ring (δ=6.8) referenced to the signal of maleic acid (δ=6.2). The content of the cyclic dimer fatty acid was successfully determined through the standard curve of mesitylene and the reported equation. Using the linearity of the mesitylene curve, the cyclic dimer fatty acid in the oil mixture was quantified. The results suggest that the proposed method can be used to quantify cyclic compounds in mixtures to optimize the dimerization process.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Mesitylene, reagent grade, 97%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Mesitylene, 98%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Lithium carbonate, 99.997% trace metals basis
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Lithium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium-7Li2 carbonate, ≥99 atom % 7Li, 99% (CP)