Merck
  • Structure and segregation of dopant-defect complexes at grain boundaries in nanocrystalline doped ceria.
Structure and segregation of dopant-defect complexes at grain boundaries in nanocrystalline doped ceria.
Physical chemistry chemical physics : PCCP (2015-05-23)
Pratik P Dholabhai, Jeffery A Aguiar, Longjia Wu, Terry G Holesinger, Toshihiro Aoki, Ricardo H R Castro, Blas P Uberuaga
ABSTRACT
Grain boundaries (GBs) dictate vital properties of nanocrystalline doped ceria. Thus, to understand and predict its properties, knowledge of the interaction between dopant-defect complexes and GBs is crucial. Here, we report atomistic simulations, corroborated with first principles calculations, elucidating the fundamental dopant-defect interactions at model GBs in gadolinium-doped and manganese-doped ceria. Gadolinium and manganese are aliovalent dopants, accommodated in ceria via a dopant-defect complex. While the behavior of isolated dopants and vacancies is expected to depend on the local atomic structure at GBs, the added structural complexity associated with dopant-defect complexes is found to have key implications on GB segregation. Compared to the grain interior, energies of different dopant-defect arrangements vary significantly at the GBs. As opposed to bulk, the stability of oxygen vacancy is found to be sensitive to the dopant arrangement at GBs. Manganese exhibits a stronger propensity for segregation to GBs than gadolinium, revealing that accommodation of dopant-defect clusters depends on the nature of dopants. Segregation strength is found to depend on the GB character, a result qualitatively supported by our experimental observations based on scanning transmission electron microscopy. The present results indicate that segregation energies, availability of favorable sites, and overall stronger binding of dopant-defect complexes would influence ionic conductivity across GBs in nanocrystalline doped ceria. Our comprehensive investigation emphasizes the critical role of dopant-defect interactions at GBs in governing functional properties in fluorite-structured ionic conductors.
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99.99% trace metals basis