Merck
  • Dietary Marine n-3 PUFAs Do Not Affect Stress-Induced Visceral Hypersensitivity in a Rat Maternal Separation Model.
Dietary Marine n-3 PUFAs Do Not Affect Stress-Induced Visceral Hypersensitivity in a Rat Maternal Separation Model.
The Journal of nutrition (2015-02-27)
Sophie A van Diest, Lieke W J van den Elsen, Allison J Klok, Olaf Welting, Francisca W Hilbers, Bert J van de Heijning, Ingrid C Gaemers, Guy E Boeckxstaens, Maria F Werner, Linette E M Willemsen, Wouter J de Jonge, René M van den Wijngaard
ABSTRACT
Although never evaluated for efficacy, n-3 (ω-3) long-chain polyunsaturated fatty acids (LCPUFAs) are commercially offered as treatment for irritable bowel syndrome (IBS). This study was designed to investigate, in a mast cell-dependent model for visceral hypersensitivity, whether this pathophysiologic mechanism can be reversed by dietary LCPUFA treatment via peroxisome proliferator-activated receptor γ (PPARG) activation. Maternally separated rats were subjected to hypersensitivity-inducing acute stress at adult age. Reversal was attempted by protocols with tuna oil-supplemented diets [4% soy oil (SO) and 3% tuna oil (SO-T3) or 3% SO and 7% tuna oil (SO-T7)] and compared with control SO diets (7% or 10% SO) 4 wk after stress. The PPARG agonist rosiglitazone was evaluated in a 1 wk preventive protocol (30 mg · kg⁻¹ · d⁻¹). Erythrocytes were assessed to confirm LCPUFA uptake and tissue expression of lipoprotein lipase and glycerol kinase as indicators of PPARG activation. Colonic mast cell degranulation was evaluated by toluidine blue staining. In vitro, human mast cell line 1 (HMC-1) cells were pretreated with rosiglitazone, eicosapentaenoic acid, or docosahexaenoic acid, stimulated with phorbol 12-myristate 13-acetate (PMA) and calcium ionophore or compound 48/80 and evaluated for tumor necrosis factor α (TNF-α) and β-hexosaminidase release. Stress led to visceral hypersensitivity in all groups. Hypersensitivity was not reversed by SO-T3 or control treatment [prestress vs. 24 h poststress vs. posttreatment area under the curve; 76 ± 4 vs. 128 ± 12 (P < 0.05) vs. 115 ± 14 and 82 ± 5 vs. 127 ± 16 (P < 0.01) vs. 113 ± 19, respectively]. Comparison of SO-T7 with its control showed similar results [74 ± 6 vs. 103 ± 13 (P < 0.05) vs. 115 ± 17 and 66 ± 3 vs. 103 ± 10 (P < 0.05) vs. 117 ± 11, respectively]. Erythrocytes showed significant LCPUFA uptake in the absence of colonic PPARG activation. Rosiglitazone induced increased PPARG target gene expression, but did not prevent hypersensitivity. Mast cell degranulation never differed between groups. Rosiglitazone and LCPUFAs significantly reduced PMA/calcium ionophore-induced TNF-α release but not degranulation of HMC-1 cells. Dietary LCPUFAs did not reverse stress-induced visceral hypersensitivity in maternally separated rats. Although further research is needed, claims concerning LCPUFAs as a treatment option in IBS cannot be confirmed at this point and should be regarded with caution.
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Dehydro-L-(+)-ascorbic acid dimer, ≥80% (enzymatic)