Merck
  • Development of a novel carbon paste sensor for determination of micromolar amounts of sulfaquinoxaline in pharmaceutical and biological samples.
Development of a novel carbon paste sensor for determination of micromolar amounts of sulfaquinoxaline in pharmaceutical and biological samples.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
Ahmad Soleymanpour, Seyyed Ahmad Rezvani
ABSTRACT
A potentiometric carbon paste sensor was fabricated for determination of sulfaquinoxaline (SQX) based on the use of ion-association complex of sulfaquinoxaline sodium with 2,3,5-triphenyltetrazolium chloride. The proposed sensor exhibited Nernstian slope of 58.4 ± 0.3 mV per decade for sulfaquinoxaline over a wide concentration range of 5.0 × 10(-6) to 1.0 × 10(-2)M, with a low detection limit of 3.0 × 10(-6)M. The sensor manifested advantages of fast response time, satisfactory reproducibility, long life time, high thermal stability and, most importantly, excellent selectivities for sulfaquinoxaline relative to a wide variety of common foreign inorganic cations, anions, sugars and amino acids. The sensor was successfully used for determination of sulfaquinoxaline in pharmaceutical solution, blood serum, urine and milk samples. The isothermal coefficient of the electrode was calculated by the investigation of temperature effects on the electrode potential response.
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Tridodecylmethylammonium chloride, 98%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Dioctyl sebacate, AldrichCPR
SAFC
Hexadecyltrimethylammonium bromide, USP/NF