Merck
  • Competency for shoot regeneration from Arabidopsis root explants is regulated by DNA methylation.
Competency for shoot regeneration from Arabidopsis root explants is regulated by DNA methylation.
Plant science : an international journal of experimental plant biology (2015-08-11)
Or Shemer, Udi Landau, Héctor Candela, Assaf Zemach, Leor Eshed Williams
ABSTRACT
Plants exhibit high capacity to regenerate in three alternative pathways: tissue repair, somatic embryogenesis and de novo organogenesis. For most plants, de novo organ initiation can be easily achieved in tissue culture by exposing explants to auxin and/or cytokinin, yet the competence to regenerate varies among species and within tissues from the same plant. In Arabidopsis, root explants incubated directly on cytokinin-rich shoot inducing medium (SIM-direct), are incapable of regenerating shoots, and a pre-incubation step on auxin-rich callus inducing medium (CIM) is required to acquire competency to regenerate on the SIM. However the mechanism underlying competency acquisition still remains elusive. Here we show that the chromomethylase 3 (cmt3) mutant which exhibits significant reduction in CHG methylation, shows high capacity to regenerate on SIM-direct and that regeneration occurs via direct organogenesis. In WT, WUSCHEL (WUS) promoter, an essential gene for shoot formation, is highly methylated, and its expression on SIM requires pre-incubation on CIM. However, in cmt3, WUS expression induced by SIM-direct. We propose that pre-incubation on CIM is required for the re-activation of cell division. Following the transfer of roots to SIM, the intensive cell division activity continues, and in the presence of cytokinin leads to a dilution in DNA methylation that allows certain genes required for shoot regeneration to respond to SIM, thereby advancing shoot formation.
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kinetin, suitable for plant cell culture, BioReagent, amorphous powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phytagel, suitable for plant cell culture, BioReagent, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Gamborg′s B-5 Basal Medium with Minimal Organics, fine powder, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O