Merck
  • Massively parallel DNA sequencing successfully identified seven families with deafness-associated MYO6 mutations: the mutational spectrum and clinical characteristics.
Massively parallel DNA sequencing successfully identified seven families with deafness-associated MYO6 mutations: the mutational spectrum and clinical characteristics.
The Annals of otology, rhinology, and laryngology (2015-05-23)
Maiko Miyagawa, Shin-Ya Nishio, Kozo Kumakawa, Shin-Ichi Usami
ABSTRACT
To elucidate the involvement of MYO6 mutations, known to be responsible for DFNA22/DFNB37, in Japanese hearing loss patients through the use of genetic analysis. Genomic variations responsible for hearing loss were identified by massively parallel DNA sequencing (MPS) of 63 target candidate genes in 1120 Japanese hearing loss patients, and the detailed clinical features for the patients with MYO6 mutations were collected and analyzed. Four mutations were successfully found in 7 families exhibiting autosomal dominant inheritance. All of the patients showed progressive hearing loss, but hearing type and onset age varied. Further, none of the affected patients showed any associated symptoms, such as hypertrophic cardiomyopathy or retinitis pigmentosa. MPS is powerful tool for the identification of rare causative deafness gene mutations, such as MYO6. The clinical characteristics noted in the present study not only confirmed the findings of previous reports but provided important new clinical information.