Merck
  • RNA steady-state defects in myotonic dystrophy are linked to nuclear exclusion of SHARP.
RNA steady-state defects in myotonic dystrophy are linked to nuclear exclusion of SHARP.
EMBO reports (2011-06-04)
Warunee Dansithong, Sonali P Jog, Sharan Paul, Robabeh Mohammadzadeh, Stephanie Tring, Yukwah Kwok, Rebecca C Fry, Paul Marjoram, Lucio Comai, Sita Reddy
ABSTRACT
We describe a new mechanism by which CTG tract expansion affects myotonic dystrophy (DM1). Changes to the levels of a panel of RNAs involved in muscle development and function that are downregulated in DM1 are due to aberrant localization of the transcription factor SHARP (SMART/HDAC1-associated repressor protein). Mislocalization of SHARP in DM1 is consistent with increased CRM1-mediated export of SHARP to the cytoplasm. A direct link between CTG repeat expression and SHARP mislocalization is demonstrated as expression of expanded CTG repeats in normal cells recapitulates cytoplasmic SHARP localization. These results demonstrate a role for the inactivation of SHARP transcription in DM1 biology.