Merck
  • Induction of cellular prion protein (PrPc) under hypoxia inhibits apoptosis caused by TRAIL treatment.
Induction of cellular prion protein (PrPc) under hypoxia inhibits apoptosis caused by TRAIL treatment.
Oncotarget (2015-03-07)
Jin-Young Park, Jae-Kyo Jeong, Ju-Hee Lee, Ji-Hong Moon, Sung-Wook Kim, You-Jin Lee, Sang-Youel Park
ABSTRACT
Hypoxia decreases cytotoxic responses to tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) protein. Cellular prion protein (PrPc) is regulated by HIF-1α in neurons. We hypothesized that PrPc is involved in hypoxia-mediated resistance to TRAIL-induced apoptosis. We found that hypoxia induced PrPc protein and inhibited TRAIL-induced apoptosis. Thus silencing of PrPc increased TRAIL-induced apoptosis under hypoxia. Overexpression of PrPc protein using an adenoviral vector inhibited TRAIL-induced apoptosis. In xenograft model in vivo, shPrPc transfected cells were more sensitive to TRAIL-induced apoptosis than in shMock transfected cells. Molecular chemo-therapy approaches based on the regulation of PrPc expression need to address anti-tumor function of TRAIL under hypoxia. Molecular chemo-therapy approaches based on the regulation of PrPc expression need to address anti-tumor function of TRAIL under hypoxia.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRNP