Merck
  • PACAP is essential for the adaptive thermogenic response of brown adipose tissue to cold exposure.
PACAP is essential for the adaptive thermogenic response of brown adipose tissue to cold exposure.
The Journal of endocrinology (2014-07-25)
Abdoulaye Diané, Nikolina Nikolic, Alexander P Rudecki, Shannon M King, Drew J Bowie, Sarah L Gray
ABSTRACT
Pituitary adenylate cyclase-activating polypeptide (PACAP) is a widely distributed neuropeptide that acts as a neurotransmitter, neuromodulator, neurotropic factor, neuroprotectant, secretagogue, and neurohormone. Owing to its pleiotropic biological actions, knockout of Pacap (Adcyap1) has been shown to induce several abnormalities in mice such as impaired thermoregulation. However, the underlying physiological and molecular mechanisms remain unclear. A previous report has shown that cold-exposed Pacap null mice cannot supply appropriate levels of norepinephrine (NE) to brown adipocytes. Therefore, we hypothesized that exogenous NE would rescue the impaired thermogenic response of Pacap null mice during cold exposure. We compared the adaptive thermogenic capacity of Pacap(-/-) to Pacap(+/+) mice in response to NE when housed at room temperature (24 °C) and after a 3.5-week cold exposure (4 °C). Biochemical parameters, expression of thermogenic genes, and morphological properties of brown adipose tissue (BAT) and white adipose tissue (WAT) were also characterized. Results showed that there was a significant effect of temperature, but no effect of genotype, on the resting metabolic rate in conscious, unrestrained mice. However, the normal cold-induced increase in the basal metabolic rate and NE-induced increase in thermogenesis were severely blunted in cold-exposed Pacap(-/-) mice. These changes were associated with altered substrate utilization, reduced β3-adrenergic receptor (β3-Ar (Adrb3)) and hormone-sensitive lipase (Hsl (Lipe)) gene expression, and increased fibroblast growth factor 2 (Fgf2) gene expression in BAT. Interestingly, Pacap(-/-) mice had depleted WAT depots, associated with upregulated uncoupling protein 1 expression in inguinal WATs. These results suggest that the impairment of adaptive thermogenesis in Pacap null mice cannot be rescued by exogenous NE perhaps in part due to decreased β3-Ar-mediated BAT activation.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Human NPPB / Natriuretic Peptides B ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Human proNPPB / Natriuretic Peptides B ELISA Kit, for serum, plasma, cell culture supernatants and urine