Merck
  • Adenovirus expressing dual c-Met-specific shRNA exhibits potent antitumor effect through autophagic cell death accompanied by senescence-like phenotypes in glioblastoma cells.
Adenovirus expressing dual c-Met-specific shRNA exhibits potent antitumor effect through autophagic cell death accompanied by senescence-like phenotypes in glioblastoma cells.
Oncotarget (2015-03-03)
Jung-Sun Lee, Eonju Oh, Ji Young Yoo, Kyeong Sook Choi, Mi Jin Yoon, Chae-Ok Yun
ABSTRACT
c-Met, a cognate receptor tyrosine kinase of hepatocyte growth factor, is overexpressed and/or mutated in number of tumors. Therefore, abrogation of c-Met signaling may serve as potential therapeutic targets. In this study, we generated Ads expressing single shRNA specific to c-Met (shMet) (dl/shMet4 and dl/shMet5) or dual shRNAs specific to c-Met (dl/shMet4+5); and examined the therapeutic potential of these newly engineered Ads in targeting c-Met, and delineated their mechanism of action in vitro and in vivo. Ads expressing shMet induced knock-down in c-Met, and phenotypically resulted in autophagy-like features including appearance of membranousvacuoles, formation of acidic vesicular organelles, and cleavage and recruitment of microtubule-associated protein1 light chain 3 to autophagosomes. Ads expressing shMet also suppressed Akt phosphorylation and increased number of senescence-related gene products including SM22, TGase II, and PAI-1. These changes resulted in inhibition of cell proliferation and G2/M arrest of U343 cells. In vivo, intratumoral injection with dl/shMet4+5 resulted in a significant reduction of tumor growth with corresponding increasing overall survival. Histopathological analysis of these treated tumors revealed that Atg5 was highly up-regulated, indicating the therapeutic induction of autophagy. In sum, these results reveal that autophagic cell death induced by shMet-expressing Ads provide a novel strategy for targeting c-Met-expressing tumors through non-apoptotic mechanism of cell death.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Acridine Orange hydrochloride solution, ≥95.0% (HPLC), 10 mg/mL in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lmna
Sigma-Aldrich
MISSION® esiRNA, targeting human LMNA
Sigma-Aldrich
DAPI, for nucleic acid staining