Merck
  • Two novel CCDC88C mutations confirm the role of DAPLE in autosomal recessive congenital hydrocephalus.
Two novel CCDC88C mutations confirm the role of DAPLE in autosomal recessive congenital hydrocephalus.
Journal of medical genetics (2012-10-09)
Anais Drielsma, Chaim Jalas, Nicolas Simonis, Julie Désir, Natalia Simanovsky, Isabelle Pirson, Orly Elpeleg, Marc Abramowicz, Simon Edvardson
ABSTRACT
Human congenital non-syndromic hydrocephalus is a vastly heterogeneous condition. A subgroup of cases are not secondary to a specific cause (eg, a neural tube defect), and within this subgroup, autosomal recessive inheritance has been described. One homozygous mutation in the DAPLE (Dvl-associating protein with a high frequency of leucine residues) protein-encoding gene CCDC88C (coiled-coil domain containing 88C) has recently been reported in a single family. The role of this gene has not been validated in another family, and no other autosomal recessive gene has been reported. We used homozygosity mapping and whole exome sequencing in two families with primary, non-syndromic congenital hydrocephalus from two different ethnic backgrounds. In each family, we identified a novel homozygous mutation of CCDC88C. One mutation produced a premature stop codon at position 312 of the protein, while the second mutation induced a frameshift in the last exon, producing a stop codon that truncated the extreme C-terminus of DAPLE, including the 2026-2028 Gly-Cys-Val motif known to bind the post synaptic density protein (PSD95), Drosophila disc large tumor suppressor (Dlg1), and zonula occludens-1 protein (zo-1) (PDZ) domain of Dishevelled. Our data validate CCDC88C as causing autosomal recessive, primary non-syndromic congenital hydrocephalus, suggesting this gene may be an important cause of congenital hydrocephalus, and underscore the important role of the C-terminal PDZ domain-binding motif in the DAPLE protein.