Merck
  • 4-(Alkylthio)- and 4-(arylthio)-benzonitrile derivatives as androgen receptor antagonists for the topical suppression of sebum production.
4-(Alkylthio)- and 4-(arylthio)-benzonitrile derivatives as androgen receptor antagonists for the topical suppression of sebum production.
Bioorganic & medicinal chemistry letters (2009-02-10)
Lorna Mitchell, Zhi Wang, Lain-Yen Hu, Catherine Kostlan, Matthew Carroll, Danielle Dettling, Daniel Du, David Pocalyko, Kimberly Wade
ABSTRACT
The first examples of thioether-substituted benzonitriles as potential soft-drug androgen receptor antagonists are reported. A number of 4-(alkylthio)- and of 4-(arylthio)-benzonitrile analogs were evaluated in human androgen receptor binding and cellular functional assays. Analogs with potent in vitro binding and cellular activities were evaluated for topical in vivo efficacy in the Golden Syrian hamster ear model. Analogs from both the 4-(alkylthio)- and of 4-(arylthio)-benzonitrile series showed moderate reduction of wax esters in vivo.