Merck
Bioreducible polymers for efficient gene and siRNA delivery.
Biomedical materials (Bristol, England) (2009-04-08)
Dhananjay Jere, Rohidas Arote, Hu-Lin Jiang, You-Kyoung Kim, Myung-Haing Cho, Chong-Su Cho
ABSTRACT
Bioreducible disulfide linkage-employing drug conjugate has already been approved for drug delivery application, and also has shown immense potential in gene and siRNA transfection. This paper will focus on the recent developments in bioreducible polymeric systems for gene and siRNA delivery application, and will discuss the advantages and challenges associated with reducible polymeric carriers.
Sigma-Aldrich
3-(Dimethylamino)-1-propylamine, purum, ≥98.0% (GC)