Merck
  • Snf2h-mediated chromatin organization and histone H1 dynamics govern cerebellar morphogenesis and neural maturation.
Snf2h-mediated chromatin organization and histone H1 dynamics govern cerebellar morphogenesis and neural maturation.
Nature communications (2014-06-21)
Matías Alvarez-Saavedra, Yves De Repentigny, Pamela S Lagali, Edupuganti V S Raghu Ram, Keqin Yan, Emile Hashem, Danton Ivanochko, Michael S Huh, Doo Yang, Alan J Mears, Matthew A M Todd, Chelsea P Corcoran, Erin A Bassett, Nicholas J A Tokarew, Juraj Kokavec, Romit Majumder, Ilya Ioshikhes, Valerie A Wallace, Rashmi Kothary, Eran Meshorer, Tomas Stopka, Arthur I Skoultchi, David J Picketts
ABSTRACT
Chromatin compaction mediates progenitor to post-mitotic cell transitions and modulates gene expression programs, yet the mechanisms are poorly defined. Snf2h and Snf2l are ATP-dependent chromatin remodelling proteins that assemble, reposition and space nucleosomes, and are robustly expressed in the brain. Here we show that mice conditionally inactivated for Snf2h in neural progenitors have reduced levels of histone H1 and H2A variants that compromise chromatin fluidity and transcriptional programs within the developing cerebellum. Disorganized chromatin limits Purkinje and granule neuron progenitor expansion, resulting in abnormal post-natal foliation, while deregulated transcriptional programs contribute to altered neural maturation, motor dysfunction and death. However, mice survive to young adulthood, in part from Snf2l compensation that restores Engrailed-1 expression. Similarly, Purkinje-specific Snf2h ablation affects chromatin ultrastructure and dendritic arborization, but alters cognitive skills rather than motor control. Our studies reveal that Snf2h controls chromatin organization and histone H1 dynamics for the establishment of gene expression programs underlying cerebellar morphogenesis and neural maturation.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys36) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Anti-Histone H3, Acetylated (1-20) Rabbit pAb, liquid, Calbiochem®