Merck
  • Loss of the B-lineage-specific gene expression program in Hodgkin and Reed-Sternberg cells of Hodgkin lymphoma.
Loss of the B-lineage-specific gene expression program in Hodgkin and Reed-Sternberg cells of Hodgkin lymphoma.
Blood (2002-10-24)
Ines Schwering, Andreas Bräuninger, Ulf Klein, Berit Jungnickel, Marianne Tinguely, Volker Diehl, Martin-Leo Hansmann, Riccardo Dalla-Favera, Klaus Rajewsky, Ralf Küppers
ABSTRACT
Hodgkin and Reed-Sternberg (HRS) cells represent the malignant cells in classical Hodgkin lymphoma (HL). Because their immunophenotype cannot be attributed to any normal cell of the hematopoietic lineage, the origin of HRS cells has been controversially discussed, but molecular studies established their derivation from germinal center B cells. In this study, gene expression profiles generated by serial analysis of gene expression (SAGE) and DNA chip microarrays from HL cell lines were compared with those of normal B-cell subsets, focusing here on the expression of B-lineage markers. This analysis revealed decreased mRNA levels for nearly all established B-lineage-specific genes. For 9 of these genes, lack of protein expression was histochemically confirmed. Down-regulation of genes affected multiple components of signaling pathways active in B cells, including B-cell receptor (BCR) signaling. Because several genes down-regulated in HRS cells are positively regulated by the transcriptional activator Pax-5, which is expressed in most HRS cells, we studied HL cell lines for mutations in the Pax-5 gene. However, no mutations were found. We propose that the lost B-lineage identity in HRS cells may explain their survival without BCR expression and reflect a fundamental defect in maintaining the B-cell differentiation state in HRS cells, which is likely caused by a novel, yet unknown, pathogenic mechanism.
Sigma-Aldrich
PAX-5 (24) Mouse Monoclonal Antibody
Sigma-Aldrich
PAX-5 (SP34) Rabbit Monoclonal Antibody