Merck
  • Suzuki coupling of aryl chlorides with phenylboronic acid in water, using microwave heating with simultaneous cooling.
Suzuki coupling of aryl chlorides with phenylboronic acid in water, using microwave heating with simultaneous cooling.
Organic letters (2005-05-20)
Riina K Arvela, Nicholas E Leadbeater
ABSTRACT
[reaction: see text]. We present here a methodology for the Suzuki coupling of aryl chlorides with phenylboronic acid using Pd/C as a catalyst, water as a solvent, and microwave heating. We show that simultaneous cooling in conjunction with microwave heating prolongs the lifetime of the aryl chloride substrates during the course of the reaction and, as a result, yields of the desired biaryl as well as overall recovery of material can be increased.
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)