Merck
  • Novel diastereoselective synthesis of bicyclic beta-lactams through radical cyclization and their reduction toward 2-(1-alkoxy-2-hydroxyethyl)piperidines and 2-(1-alkoxy-2-hydroxyethyl)azepanes.
Novel diastereoselective synthesis of bicyclic beta-lactams through radical cyclization and their reduction toward 2-(1-alkoxy-2-hydroxyethyl)piperidines and 2-(1-alkoxy-2-hydroxyethyl)azepanes.
The Journal of organic chemistry (2008-01-24)
Erika Leemans, Matthias D'hooghe, Yves Dejaegher, Karl W Törnroos, Norbert De Kimpe
ABSTRACT
1-Allyl- and 1-(3-phenylallyl)-substituted 4-(2-bromo-1,1-dimethylethyl)azetidin-2-ones were transformed into 3-substituted 7-alkoxy-5,5-dimethyl-1-azabicyclo[4.2.0]octane-8-ones through radical cyclization by means of n-tributyltin hydride and AIBN in toluene with excellent diastereocontrol (>or=99%). The radical cyclization of 4-(2-bromo-1,1-dimethylethyl)-1-(2-methylallyl)azetidin-2-ones afforded 8-alkoxy-3,6,6-trimethyl-1-azabicyclo[5.2.0]nonan-9-ones in good diastereomeric excess (75-78%). The reductive ring opening of 1-azabicyclo[4.2.0]octane-8-ones and 1-azabicyclo[5.2.0]nonan-9-ones with lithium aluminum hydride resulted in novel 2-(1-alkoxy-2-hydroxyethyl)piperidines and -azepanes, which were isolated as single isomers.
Sigma-Aldrich
3-Bromo-2,2-dimethyl-1-propanol, 96%