Merck
[Acyclic nucleotide analogues based on phosphonic acids].
Bioorganicheskaia khimiia (1998-04-29)
E V Efimtseva, S N Mikhaĭlov, M V Fomicheva, S V Meshkov, M S Rodionov, A R Khomutov, E De Clercq
ABSTRACT
The synthesis of novel nucleotide analogues on the basis of ethylphosphonic acid was described. A rigid structural element, an amide or a double bond, was characteristics of the compounds synthesized. The antiviral and cytotoxic activities of these compounds were studied in cell cultures.
Sigma-Aldrich
Ethylphosphonic acid, 98%