Merck
  • Room temperature copper(II)-catalyzed oxidative cyclization of enamides to 2,5-disubstituted oxazoles via vinylic C-H functionalization.
Room temperature copper(II)-catalyzed oxidative cyclization of enamides to 2,5-disubstituted oxazoles via vinylic C-H functionalization.
The Journal of organic chemistry (2012-07-31)
Chi Wai Cheung, Stephen L Buchwald
ABSTRACT
A copper(II)-catalyzed oxidative cyclization of enamides to oxazoles via vinylic C-H bond functionalization at room temperature is described. Various 2,5-disubstituted oxazoles bearing aryl, vinyl, alkyl, and heteroaryl substituents could be synthesized in moderate to high yields. This reaction protocol is complementary to our previously reported iodine-mediated cyclization of enamides to afford 2,4,5-trisubstituted oxazoles.