Merck
  • No conclusive evidence that histologically found granulomas and acute local reactions following hylan G-F 20 injections are related or have clinical significance.
No conclusive evidence that histologically found granulomas and acute local reactions following hylan G-F 20 injections are related or have clinical significance.
The journal of knee surgery (2013-07-23)
David D Waddell, Alan Beyer, Terry L Thompson, Jennifer Morawiak, Clare Elkins, Andrew Rosenberg, Andrew Spitzer
ABSTRACT
Osteoarthritic (OA) knee pain can be successfully treated with hylan G-F 20 (Synvisc, Sanofi Biosurgery, Cambridge, MA) with few local adverse events. However, a few studies have identified hyaluronate positive (HA+) granulomas in the synovial tissue of patients treated with hylan G-F 20 raising the question of their relationship and clinical significance. To understand the potential relationship of HA+ granulomas with the occurrence of acute local reactions (ALRs), we evaluated the synovial tissue of OA patients undergoing total knee replacement that had previously been treated with hylan G-F 20 (n = 101) or had not been treated (n = 20). Granulomas were observed in nine patients, of which eight were in the hylan G-F 20 group (7.9%); HA+ granulomas were identified in six of these eight patients (5.9%). Three of the six patients with HA+ granulomas experienced an ALR within 30 days of administering an injection. Overall, we found no consistent relationship between histologically found HA+ granulomas and the occurrence of an ALR following hylan G-F 20 treatment. These microscopic granulomas were not associated with any symptoms and likely have little clinical significance. The low occurrence of granulomas and/or ALRs should not preclude use of hylan G-F 20 for the treatment of knee pain associated with OA.
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,750,000-2,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor