Merck
  • L-arginine chlorination products inhibit endothelial nitric oxide production.
L-arginine chlorination products inhibit endothelial nitric oxide production.
The Journal of biological chemistry (2001-05-26)
C Zhang, C Reiter, J P Eiserich, B Boersma, D A Parks, J S Beckman, S Barnes, M Kirk, S Baldus, V M Darley-Usmar, C R White
ABSTRACT
The myeloperoxidase-derived oxidant hypochlorous acid (HOCl) is thought to contribute to endothelial dysfunction, but the mechanisms underlying this inhibitory effect are unknown. The present study tested the hypothesis that HOCl and L-arginine (L-Arg) react to form novel compounds that adversely affect endothelial function by inhibiting nitric oxide (NO) formation. Using spectrophotometric techniques, we found that HOCl and L-Arg react rapidly (k = 7.1 x 10(5) m(-1) s(-1)) to form two major products that were identified by mass spectrometry as monochlorinated and dichlorinated adducts of L-Arg. Pretreatment of bovine aortic endothelial cells with the chlorinated L-Arg metabolites (Cl-l-Arg) inhibited the -induced formation of the NO metabolites nitrate (NO(3)(-)) and nitrite (NO(2)(-)) in a concentration-dependent manner. Preincubation of rat aortic ring segments with Cl-L-Arg resulted in concentration-dependent inhibition of acetylcholine-induced relaxation. In contrast, blood vessels relaxed normally to the endothelium-independent vasodilator sodium nitroprusside. In vivo administration of Cl-L-Arg to anesthetized rats increased carotid artery vascular resistance. A greater than 10-fold excess of L-Arg was required to reverse the inhibitory effects of Cl-L-Arg in vivo and in vitro. Reaction of HOCl with D-arginine (D-Arg) did not result in the formation of inhibitory products. These results suggest that HOCl reacts with L-Arg to form chlorinated products that act as nitric-oxide synthase inhibitors.
Product Description
Supelco
Diaion® CR11 sodium form, pkg of 1000 g