Merck
  • Molecular layer deposition of functional thin films for advanced lithographic patterning.
Molecular layer deposition of functional thin films for advanced lithographic patterning.
ACS applied materials & interfaces (2011-02-10)
Han Zhou, Stacey F Bent
ABSTRACT
Photoresist materials comprise one of the main challenges faced by lithography to meet the requirements of electronic device size scaling. Here we report for the first time the use of molecular layer deposition (MLD) to produce photoresist materials with controllable placement of functional moieties. Polyurea resists films are deposited by MLD using urea coupling reactions between 1,4-phenylene diisocyanate (PDIC) and ethylenediamine (ED) or 2,2'-(propane-2,2-diylbis(oxy))diethanamine (PDDE) monomers in a layer-by-layer fashion with a linear growth rate, allowing acid-labile groups to be incorporated into the film at well-controlled positions. The films are deposited with stoichiometric compositions and have highly uniform surface morphology as investigated using atomic force microscopy. We show that acid treatment can cleave the backbone of the polyurea film at positions where the acid-labile groups are embedded. We further show that after soaking the polyurea film with photoacid generator (PAG), it acts as a photoresist material and we present several UV patterning demonstrations. This approach presents a new way to make molecularly designed resist films for lithography.
Sigma-Aldrich
1,4-Phenylene diisocyanate