Merck
  • A catalytic one-step process for the chemo- and regioselective acylation of monosaccharides.
A catalytic one-step process for the chemo- and regioselective acylation of monosaccharides.
Journal of the American Chemical Society (2007-10-02)
Takeo Kawabata, Wataru Muramatsu, Tadashi Nishio, Takeshi Shibata, Hartmut Schedel
ABSTRACT
An organocatalytic method for the chemo- and regioselective acylation of monosaccharides has been developed. Treatment of octyl beta-D-glucopyranoside with isobutyric anhydride in the presence of 10 mol % of a C2-symmetric chiral 4-pyrrolidinopyridine catalyst (1) at -50 degrees C gave the 4-O-isobutyryl derivative as the sole product in 98% yield. Thus, chemoselective acylation, favoring a secondary hydroxyl group in the presence of a free primary hydroxyl group, and regioselective acylation, favoring one of three secondary hydroxyl groups, took place with perfect selectivity. A competitive acylation between octyl beta-D-glucopyranoside and a primary alcohol (2-phenylethanol) with 1.1 equiv of isobutyric anhydride in the presence of 1 gave the 4-O-isobutyrate of octyl beta-D-glucopyranoside with 99% regioselectivity in 98% yield, which indicates that acylation of the secondary hydroxyl group at C(4) of the carbohydrate proceeds in an accelerative manner. A possible mechanism, involving multiple hydrogen-bonding between 1 and the monosaccharide, is proposed for the chemo- and regioselective acylation.
Sigma-Aldrich
Isobutyric anhydride, 97%