Merck
Synthesis and properties of supramolecular ionic networks.
Journal of the American Chemical Society (2008-07-03)
Michel Wathier, Mark W Grinstaff
ABSTRACT
The synthesis of a supramolecular ionic network, its physical properties, and the use of this network property to form macroscopic porphyrin fibers are described. These ionic networks are compared to ionic liquids. Current ionic liquid compositions have a charge and molar ratio of 1:1 where an anionic species is matched with a cationic species; however, alteration of this molar ratio while maintaining the charge ratio of 1:1 by using multivalent cationic/anionic molecular pairs affords new ionically cross-linked networks with interesting properties.