Merck
  • Metabolism and metabolic effects of 2-azahypoxanthine and 2-azaadenosine.
Metabolism and metabolic effects of 2-azahypoxanthine and 2-azaadenosine.
Biochemical pharmacology (1985-04-15)
L L Bennett, D Smithers, L M Rose, D J Adamson, S C Shaddix, H J Thomas
ABSTRACT
The metabolism and metabolic effects of 2-azahypoxanthine and 2-azaadenosine were studied to elucidate the biochemical basis for their known cytotoxicities. 2-Azaadenosine is a known substrate for adenosine kinase. That 2-azahypoxanthine is a substrate for hypoxanthine (guanine) phosphoribosyltransferase is shown by the observations that, in cell-free fractions from HEp-2 cells supplemented with 5-phosphoribosyl-1-pyrophosphate, 2-azahypoxanthine inhibited the conversion of hypoxanthine to IMP but not the conversion of adenine to AMP, and hypoxanthine, but not adenine, inhibited the conversion of 2-azahypoxanthine to 2-azaIMP. [8-14C]2-Azahypoxanthine was synthesized from [8-14C]hypoxanthine via [2-14C]-4-amino-5-imidazolecarboxamide. In HEp-2 cells in culture, the principal metabolite of [8-14C]-2-azahypoxanthine was 2-azaATP; there was no detectable 14C in deoxynucleotides or in DNA or RNA fractions. 2-Azaadenosine was much more toxic than 2-azahypoxanthine, and, when used in the presence of an adenosine deaminase inhibitor, 2'-deoxycoformycin, was converted in HEp-2 cells to 2-azaATP in amounts that exceeded those of ATP in control cells. The pool of ATP was reduced by as much as 75% as 2-azaATP accumulated. In a short-term experiment (4 hr), 2-azaadenosine selectively reduced the pools of adenine nucleotides, whereas 2-azahypoxanthine reduced the pools of guanine nucleotides selectively. Both 2-azahypoxanthine and 2-azaadenosine inhibited the incorporation of formate into purine nucleotides and were without effect on the conversion of thymidine and uridine to nucleotides. 2-Azahypoxanthine inhibited the incorporation of thymidine into macro-molecules but not that of uridine or leucine; 2-azaadenosine inhibited the incorporation of all three of these precursors non-selectively. 2-AzaIMP inhibited IMP dehydrogenase competitively with IMP (Ki = 66 microM). The difference in effects of 2-azahypoxanthine and 2-azaadenosine perhaps may be due to the production, from 2-azahypoxanthine but not from 2-azaadenosine + 2'-deoxycoformycin, of 2-azaIMP, which inhibits synthesis of guanine nucleotides and thereby results in inhibition of DNA synthesis. Specific sites of action for 2-azaadenosine are yet undefined.
Product Description
Dacarbazine impurity A, European Pharmacopoeia (EP) Reference Standard