Merck
  • Non-invasive analysis of penetration and storage of Isoconazole nitrate in the stratum corneum and the hair follicles.
Non-invasive analysis of penetration and storage of Isoconazole nitrate in the stratum corneum and the hair follicles.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2012-01-31)
J Lademann, A Patzelt, S Schanzer, H Richter, G Thiede, B Havlickova, C Günther, M Friedrich, W Sterry, J W Fluhr, S Seifert
ABSTRACT
Bacteria and fungi are located in the stratum corneum and the hair follicles. Therefore, the development and assessment of efficient drugs requires standard in vivo investigation methods permitting a differentiation between intercellular and follicular penetration and storage of topically applied anti-microbial substances. In the present study, the penetration and storage of Isoconazole nitrate in the stratum corneum and hair follicles was investigated by differential stripping after a 14-day topical application period and during a follow-up period of a further 21 days. One week after the application had terminated, Isoconazole nitrate could still be detected in concentrations above the minimal inhibition concentration in the stratum corneum and the hair follicles. In some subjects, Isoconazole nitrate could even be detected 14 days after the last application. No relevant changes in TEWL values were measured, indicating that the investigated compound did not induce an impairment of the barrier function. The study showed that differential stripping is suited to investigate the penetration and storage of topically applied substances into the stratum corneum and the hair follicles. Also, the hair follicles are a long-term reservoir for topically applied substances. This is of clinical importance, where a long-lasting therapeutic effect beyond the application time is required.
Product Description
Isoconazole, European Pharmacopoeia (EP) Reference Standard
Isoconazole nitrate, European Pharmacopoeia (EP) Reference Standard