- Isolation and structural elucidation of flibanserin as an adulterant in a health supplement used for female sexual performance enhancement.
Isolation and structural elucidation of flibanserin as an adulterant in a health supplement used for female sexual performance enhancement.
Journal of pharmaceutical and biomedical analysis (2011-10-01)
Min-Yong Low, Lin Li, Xiaowei Ge, Chee-Leong Kee, Hwee-Ling Koh
PMID21955644
ABSTRACT
A health supplement used for female sexual performance enhancement was sent to Health Sciences Authority of Singapore for testing. An unknown compound was detected and isolated from the health supplement and its structure was elucidated using LC-DAD, LC-FTMS, NMR and IR. The detected compound was identified to be flibanserin, a non-hormonal treatment developed for pre-menopausal woman with hypoactive sexual desire disorder (HSDD).
MATERIALS