Merck
  • Antioxidant and enzyme inhibition activities and chemical profiles of Polygonum sachalinensis F.Schmidt ex Maxim (Polygonaceae).
Antioxidant and enzyme inhibition activities and chemical profiles of Polygonum sachalinensis F.Schmidt ex Maxim (Polygonaceae).
Fitoterapia (2009-08-25)
Peihong Fan, Lara Terrier, Anne-Emmanuelle Hay, Andrew Marston, Kurt Hostettmann
ABSTRACT
Polygonum sachalinensis is a widespread invasive plant in Europe. Chemical profiles of its different organs were studied by HPLC-UV-ESI/MS. Seven major constituents quercetin-3-O-beta-D-galactopyranoside, quercetin-3-O-arabinopyranoside, lapathoside D, N-trans-feruloyltyramine, lapathoside C, hydropiperoside, and vanicoside B were isolated and identified. The free radical-scavenging, alpha/beta-glucosidase, and acetylcholinesterase inhibitory activities of crude MeOH extracts and isolated compounds were studied. The structure-activity relationships were discussed. The chemical profiles revealed flavonoids and phenylpropanoids are the major compounds of all the organs of this plant. Quercetin-3-O-arabinopyranoside, lapathoside D, N-trans-feruloyltyramine, lapathoside C and hydropiperoside were isolated from this species for the first time. In the alpha-glucosidase bioassay, quercetin-3-O-beta-D-galactopyranoside, lapathoside D and N-trans-feruloyltyramine demonstrated stronger activities than the positive reference acarbose. The trend in scavenging power showed no relation to enzyme inhibition in the test models.
Sigma-Aldrich
Moupinamide, ≥95% (LC/MS-ELSD)