- Effect of high pressure on isomerization and degradation of lactose in alkaline media.
Effect of high pressure on isomerization and degradation of lactose in alkaline media.
Journal of agricultural and food chemistry (2003-03-20)
F Javier Moreno, Mar Villamiel, Agustín Olano
PMID12643648
ABSTRACT
The combined effects of temperature (60 degrees C) and high-pressure treatments (400 MPa for 3 h) on the isomerization-degradation of lactose (10%) in basic media were studied. The formation of isomeric disaccharides (lactulose and epilactose) and galactose decreased by the application of high pressure in aqueous sodium hydroxide 4 mM (pH 10.2) and 8 mM (pH 10.6), and sodium carbonate-bicarbonate buffer (pH 10.0). In addition, no substantial color development was observed in the sodium hydroxide systems for high and atmospheric pressure, whereas the application of high pressure led to a noticeable decrease of color development in the carbonate-bicarbonate system.