Merck
  • Application of gradient-enhanced nuclear Overhauser effect spectroscopy GOESY) in the structure elucidation of plant secondary metabolites.
Application of gradient-enhanced nuclear Overhauser effect spectroscopy GOESY) in the structure elucidation of plant secondary metabolites.
Phytochemical analysis : PCA (2001-11-14)
S D Sarker, Z Latif, R J Nash
ABSTRACT
Gradient enhanced nuclear Overhauser effect spectroscopy (GOESY) has been applied in the structural elucidation of five plant secondary metabolites: eucalyptin, arctigenin, 5-geranyloxy-7-methoxycoumarin, 2,6-dihydroxy-4-methoxyisovalerophenone and N-feruloyltyramine. The importance and sensitivity of this technique in structure elucidation of plant secondary metabolites are discussed.
Sigma-Aldrich
Moupinamide, ≥95% (LC/MS-ELSD)