Merck
The major flavonoid of Dodonaea angustifolia.
Fitoterapia (2001-07-14)
F R van Heerden, A M Viljoen, B E van Wyk
ABSTRACT
The major leaf flavonoid of Dodonaea angustifolia, an important South African traditional medicine, has been identified as 5,7,4'-trihydroxy-3,6-dimethoxyflavone (1).
Sigma-Aldrich
4′,5,7-Trihydroxy-3,6-dimethoxyflavone, ≥90% (LC/MS-ELSD)