Merck
  • ESR evidence for superoxide, hydroxyl radicals and singlet oxygen produced from hydrogen peroxide and nickel(II) complex of glycylglycyl-L-histidine.
ESR evidence for superoxide, hydroxyl radicals and singlet oxygen produced from hydrogen peroxide and nickel(II) complex of glycylglycyl-L-histidine.
Biochemical and biophysical research communications (1989-03-15)
S Inoue, S Kawanishi
ABSTRACT
ESR studies using spin traps, 5,5-dimethylpyrroline-N-oxide and alpha-(4-pyridyl 1-oxide)-N-tert-butylnitrone, revealed that hydroxyl radical adducts are produced by the decomposition of hydrogen peroxide in the presence of nickel(II) oligopeptides. Order of catalytic activities of nickel(II) oligopeptides used in the production of hydroxyl radical adducts was tetraglycine greater than pentaglycine greater than triglycine greater than GlyGly, GlyHis. Ni(II) GlyGlyHis plus hydrogen peroxide produced superoxide in addition to hydroxyl radical adduct. Trapping experiments with 2,2,6,6-tetramethyl-4-piperidone suggested that singlet oxygen was generated by the reaction of hydrogen peroxide with Ni(II) GlyGlyHis, but not in the case of tetraglycine, pentaglycine, triglycine, GlyGly or GlyHis.
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidone, 95%
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, 99%