Merck
  • A highly efficient and aerobic protocol for the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives via a palladium-catalyzed ligand-free Suzuki reaction.
A highly efficient and aerobic protocol for the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives via a palladium-catalyzed ligand-free Suzuki reaction.
Organic & biomolecular chemistry (2012-08-15)
Xiaofeng Rao, Chun Liu, Jieshan Qiu, Zilin Jin
ABSTRACT
A palladium-catalyzed aerobic and ligand-free Suzuki reaction in aqueous ethanol has been developed for the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives. A number of N-heteroaryl halides, namely 2-halogenated pyridines, 2-bromoquinoline, 5-bromopyrimidine and 2-chloropyrazine, were coupled with 4-(9H-carbazol-9-yl)phenylboronic acid (CPBA) or 9-phenyl-9H-carbazol-3-ylboronic acid (PCBA) efficiently to afford good to excellent yields in a short reaction time. Moreover, the catalytic system of Pd(OAc)(2)-EtOH/H(2)O-K(2)CO(3) was successfully extended to the cross-couplings of N-heteroaryl halides with various arylboronic acids. The results demonstrated that the cross-coupling reaction in the present protocol was promoted by oxygen.
Sigma-Aldrich
5-Bromopyrimidine, 97%