Merck
  • Cloning of nicotianamine synthase genes, novel genes involved in the biosynthesis of phytosiderophores.
Cloning of nicotianamine synthase genes, novel genes involved in the biosynthesis of phytosiderophores.
Plant physiology (1999-02-10)
K Higuchi, K Suzuki, H Nakanishi, H Yamaguchi, N K Nishizawa, S Mori
ABSTRACT
Nicotianamine synthase (NAS), the key enzyme in the biosynthetic pathway for the mugineic acid family of phytosiderophores, catalyzes the trimerization of S-adenosylmethionine to form one molecule of nicotianamine. We purified NAS protein and isolated the genes nas1, nas2, nas3, nas4, nas5-1, nas5-2, and nas6, which encode NAS and NAS-like proteins from Fe-deficient barley (Hordeum vulgare L. cv Ehimehadaka no. 1) roots. Escherichia coli expressing nas1 showed NAS activity, confirming that this gene encodes a functional NAS. Expression of nas genes as determined by northern-blot analysis was induced by Fe deficiency and was root specific. The NAS genes form a multigene family in the barley and rice genomes.
Product Description
ProbeQuant G-50 Micro Columns, Cytiva 28-9034-08, pack of 50 columns