Merck
  • Variability and corresponding amplitude-velocity relation of activity propagating in one-dimensional neural cultures.
Variability and corresponding amplitude-velocity relation of activity propagating in one-dimensional neural cultures.
Journal of neurophysiology (2007-03-09)
Shimshon Jacobi, Elisha Moses
ABSTRACT
We investigate the propagation of neural activity along one-dimensional rat hippocampal cultures patterned in lines over multielectrode arrays. Activity occurs spontaneously or is evoked by local electrical or chemical stimuli, with different resulting propagation velocities and firing rate amplitudes. A variability of an order of magnitude in velocity and amplitude is observed in spontaneous activity. A linear relation between velocity and amplitude is identified. We define a measure for neuron activation synchrony and find that it correlates with front velocity and is higher for electrically evoked fronts. We present a model that explains the linear relation between amplitude and velocity, which highlights the role of synchrony. The relation to current models for signal propagation in neural media is discussed.
Sigma-Aldrich
SODOSIL RA 14, mildly alkaline, chlorine and phosphate-free cleaner
Sigma-Aldrich
SODOSIL RA 13, highly alkaline cleaner