Merck
  • In Vitro Antileishmanial Activity of Essential Oil of Vanillosmopsis arborea (Asteraceae) Baker.
In Vitro Antileishmanial Activity of Essential Oil of Vanillosmopsis arborea (Asteraceae) Baker.
Evidence-based complementary and alternative medicine : eCAM (2013-08-13)
Aracélio Viana Colares, Fernando Almeida-Souza, Noemi Nosomi Taniwaki, Celeste da Silva Freitas Souza, José Galberto Martins da Costa, Kátia da Silva Calabrese, Ana Lúcia Abreu-Silva
ABSTRACT
The search for new immunopharmacological chemical agents to treat various diseases caused by bacteria, fungi, and protozoa, such as leishmaniasis, for example, has led to the exploration of potential products from plant species and their main active ingredients. Antimonial drugs are the current treatment for leishmaniasis. These drugs cause major side effects and frequent discontinuation of treatment. In this study, we evaluated the in vitro leishmanicidal activity of essential oil of Vanillosmopsis arborea (VAEO) and its major compound α -bisabolol against Leishmania amazonensis. The essential oil and α -bisabolol showed activity against promastigotes (IC50 7.35 and 4.95  μ g/mL resp.) and intracellular amastigotes (IC50 12.58 and 10.70  μ g/mL, resp.). Neither product showed any cytotoxicity on treated macrophages. The ultrastructural analysis of promastigotes incubated with VAEO or α -bisabolol at 30  μ g/mL, showed morphological changes with the accumulation of vesicles electrodense lipid inclusions. The results give evidence that both VAEO and α -bisabolol have potential as new therapeutic agents against leishmaniasis.
Sigma-Aldrich
(−)-α-Bisabolol, ≥93% (GC)