Merck
2-Amino-5-bromo-pyridine-benzoic acid (1/1).
Acta crystallographica. Section E, Structure reports online (2010-01-01)
Madhukar Hemamalini, Hoong-Kun Fun
ABSTRACT
In the title adduct, C(5)H(5)BrN(2)·C(7)H(6)O(2), the carboxyl group of the benzoic acid mol-ecule is twisted away from the attached ring by 12.97 (11)°. The 2-amino-5-bromo-pyridine mol-ecules inter-act with the carboxylic group of neighbouring benzoic acid mol-ecules through N-H⋯O and O-H⋯N hydrogen bonds, forming cyclic R(2) (2)(8) hydrogen-bonded motifs and linking the mol-ecules into a two-dimensional network lying parallel to (100). The crystal structure is further stabilized by weak C-H⋯O hydrogen bonds.
Sigma-Aldrich
2-Amino-5-bromopyridine, 97%