Merck
Vitamin K concentrations in the plasma and liver of surgical patients.
The American journal of clinical nutrition (1990-05-01)
Y Usui, H Tanimura, N Nishimura, N Kobayashi, T Okanoue, K Ozawa
ABSTRACT
Vitamin K deficiency has been reported in patients who were treated with antibiotics and placed on poor diets after surgery. High-performance liquid chromatography (HPLC) was used to study the influence of dietary intake on vitamin K concentrations in surgical patients (n = 22). Plasma phylloquinone decreased rapidly from 1.19 +/- 0.16 to 0.47 +/- 0.12 nmol/L (means +/- SEM, n = 11) on a low-phylloquinone diet and from 1.16 +/- 0.12 to 0.36 +/- 0.07 nmol/L (n = 11) by postoperative fasting. A small amount of phylloquinone and a large amount of menaquinone were found in liver tissue. Phylloquinone concentration was 28.0 +/- 4.3 pmol/g liver (wet weight) on the standard diet (n = 7) whereas it was 6.8 +/- 1.1 pmol/g on the low-phylloquinone diet after 3 d (n = 8). Because phylloquinone is rapidly depleted by fasting, it may be difficult to prevent vitamin K deficiency by dietary phylloquinone alone during long-term fasting after surgery.
Sigma-Aldrich
Menadione, meets USP testing specifications
Supelco
Menadione (K3), analytical standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Menadione, crystalline