Merck
  • Simultaneous determination of methionine sulfoxide and methionine in blood plasma using gas chromatography-mass spectrometry.
Simultaneous determination of methionine sulfoxide and methionine in blood plasma using gas chromatography-mass spectrometry.
Analytical biochemistry (2003-02-11)
Ryuichi Mashima, Takako Nakanishi-Ueda, Yorihiro Yamamoto
ABSTRACT
Methionine sulfoxide is an oxidation product of methionine with reactive oxygen species via 2-electron-dependent mechanism. Such oxidants can be generated from activated neutrophils; therefore, methionine sulfoxide can be regarded as a biomarker of oxidative stress in vivo. We describe here a method for the simultaneous determination of methionine sulfoxide and methionine in blood plasma using gas chromatography-mass spectrometry with isotopically labeled compounds as internal standards. This method comprises the inclusion of [Me-13C, Me-2H(3)]methionine sulfoxide and [Me-13C, Me-2H(3)]methionine into plasma, the removal of plasma proteins using acetonitrile, the purification of amino acids with cation-exchange chromatography, and the derivatization of methionine sulfoxide and methionine to their corresponding tert-butyldimethylsilyl derivatives using N-(tert-butyldimethylsilyl)-N-methyltrifluoroacetamide. Quantitation was performed by electron impact mode. The levels of methionine sulfoxide in healthy human blood plasma were 4.0 +/- 1.0 microM (means +/- SD, n = 8), indicating that approximately 10% of methionine is detected as the oxidized form in healthy human plasma. The ratio of methionine sulfoxide in total methionine increased with treatment of human blood with phorbol 12-myristate 13-acetate, while this ratio remained constant in plasma from alloxan-induced hyperglycemic rabbits. These results indicate that this method is applicable for plasma samples and methionine sulfoxide can represent oxidative stress caused by nonradical oxidation in vivo.
Product Description
Supelco
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, derivatization grade (GC derivatization), LiChropur, ≥98.0% (GC)
Sigma-Aldrich
DL-Methionine sulfoxide, ≥98.5% (NT)