Merck
  • Acute hepatitis caused by alverine associated with anti-lamin A and C autoantibodies.
Acute hepatitis caused by alverine associated with anti-lamin A and C autoantibodies.
Journal of hepatology (1997-08-01)
D Malka, B N Pham, J C Courvalin, M Corbic, D Pessayre, S Erlinger
ABSTRACT
We report the case of a 67-year-old woman in whom onset and regression of acute hepatitis were closely related to the time of administration and withdrawal of the smooth muscle relaxant alverine. Antinuclear antibodies were positive, and their titer followed the course of hepatitis. They presented a smooth rim-like nuclear immunofluorescence staining pattern. Immunoblot assay showed that they were directed against lamin A and lamin C. This suggests that alverine should be added to the list of drugs known to produce acute hepatitis, and that drug-induced liver injury is a possible cause of antinuclear antibodies specific for lamin A and lamin C.
Product Description
Alverine citrate salt, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Alverine citrate salt