Merck
  • Distribution, prevalence, and drug binding profile of gamma-aminobutyric acid type A receptor subtypes differing in the beta-subunit variant.
Distribution, prevalence, and drug binding profile of gamma-aminobutyric acid type A receptor subtypes differing in the beta-subunit variant.
The Journal of biological chemistry (1994-10-28)
D Benke, J M Fritschy, A Trzeciak, W Bannwarth, H Mohler
ABSTRACT
Native gamma-aminobutyric acid type A (GABAA) receptors containing different beta-subunit variants were identified immunobiochemically with antisera recognizing selectively the beta 1-, beta 2-, or beta 3-subunit. As determined by immunoprecipitation, the beta 2-subunit was present in 55-60% of GABAA receptors, while only minor receptor populations contained the beta 1-subunit (16-18%) or the beta 3-subunit (19-25%). Since the sum of these values amounts to about 100%, it is concluded that GABAA receptors largely contain only a single type of beta-subunit. Pharmacologically, receptors containing the beta 2-subunit differed from those containing the beta 1- or beta 3-subunit by their differential affinities for benzodiazepine receptor ligands. The subunit composition was analyzed biochemically in receptors immunoprecipitated by the beta 2-subunit antiserum. The beta 2-subunit was preferentially associated with the alpha 1-subunit (rarely with the alpha 2-subunit) and with the gamma 2-subunit; negligible or no immunoreactivity was detected for the alpha 3-, alpha 5-, or beta 1-subunit. A stringent co-expression of alpha 1- and beta 2-subunits was confirmed by double immunofluorescence staining on the cellular level. Neurons expressing the beta 3-subunit immunoreactivity were largely double labeled by the alpha 2-subunit antiserum. Thus, the subunit combinations alpha 1 beta 2 gamma 2 and alpha 2 beta 3 gamma 2 represent two main GABAA receptor subtypes, which together amount to 75-85% of the diazepam-sensitive GABAA receptors.