Merck
  • Metabolism of ethylmalonate to mesaconate in the rat. Evidence for trans-dehydrogenation of methylsuccinate.
Metabolism of ethylmalonate to mesaconate in the rat. Evidence for trans-dehydrogenation of methylsuccinate.
The Biochemical journal (1983-08-15)
J A Montgomery, O A Mamer, C R Scriver
ABSTRACT
Rats excrete increased ethylmalonate and methylsuccinate when given ethylmalonate in the diet; when given methylsuccinate they excrete methylsuccinate and mesaconate. Tenfold more labelled mesaconate was produced from threo-methyl[2,3-2H2] succinate precursor than from the erythro isomer. Our findings suggest trans-dehydrogenation of methylsuccinate and are the first direct evidence linking the metabolism of ethylmalonate, methylsuccinate and mesaconate.
Sigma-Aldrich
Methylsuccinic acid, 99%