Merck
  • Ultrastructure of the choroid plexus epithelium of pigeons treated with drugs: I. Effect of Thyradin, 2,4-dinitrophenol, and cycloheximide.
Ultrastructure of the choroid plexus epithelium of pigeons treated with drugs: I. Effect of Thyradin, 2,4-dinitrophenol, and cycloheximide.
The American journal of anatomy (1987-02-01)
K Koshiba
ABSTRACT
Possible changes in the epithelial cells of the pigeon choroid plexus induced by administration of thyroid powder (Thyradin), 2,4-dinitrophenol, and cycloheximide were studied by scanning and transmission electron microscopy. A marked increase in the number of large bulbous and bleblike protrusions on the apical end of the epithelial cells was observed after oral administration of Thyradin for a month. The endoplasmic content of the protrusions consisted mainly of electron-lucent material. These results provide morphological evidence for the stimulatory effect of Thyradin. Intramuscular injections of 2,4-dinitrophenol for 15 days caused the collapse or deformation of the mitochondria and bleblike or bulbous protrusions. This indicates that changes in the surface configuration of the choroid plexus are controlled by an energy-dependent mechanism. The decrease of protrusions and polyribosomes and increase of the tubular saccules of varying electron density, size, and shape were noted in cells after 15 days of intramuscular cycloheximide injection. The electron density of the protrusions is lower than that of the control pigeons. The results of this study suggest that a curious pleomorphic structure on the apical surface of the choroid epithelial cell of pigeon is closely related to the functional state of choroidal cells. The study also demonstrates that a secondary ultrastructural response due to diverse physiologic effects is reflected in the architecture of the choroid plexus cells.