Merck
  • Variation in diagnostic performance among radiologists at screening CT colonography.
Variation in diagnostic performance among radiologists at screening CT colonography.
Radiology (2013-03-02)
B Dustin Pooler, David H Kim, Cesare Hassan, Antonio Rinaldi, Elizabeth S Burnside, Perry J Pickhardt
ABSTRACT
To assess the variation in diagnostic performance among radiologists at screening computed tomographic (CT) colonography. In this HIPAA-compliant, institutional review board-approved study, 6866 asymptomatic adults underwent first-time CT colonographic screening at a single center between January 2005 and November 2011. Results of examinations were interpreted by one of eight board-certified abdominal radiologists (mean number of CT colonographic studies per reader, 858; range, 131-2202). Findings at CT colonography and subsequent colonoscopy were recorded, and key measures of diagnostic performance, including adenoma and advanced neoplasia detection rate, were compared among the radiologists. The overall prevalence of histopathologically confirmed advanced neoplasia was 3.6% and did not differ significantly among radiologists (range, 2.4%-4.4%; P = .067; P = .395 when one outlier was excluded). Overall, 19.5% of polyps detected at CT colonography proved to be advanced neoplasia and did not differ significantly among radiologists (range, 14.4%-23.2%; P = .223). The overall per-polyp endoscopic confirmation rate was 93.5%, ranging from 80.0% to 97.6% among radiologists (P = .585). The overall percentage of nondiagnostic CT colonographic examinations was 0.7% and was consistent among radiologists (range, 0.3%-1.1%; P = .509). Consistent performance for adenoma and advanced neoplasia detection, as well as other clinically relevant end points, were observed among radiologists at CT colonographic screening.
Sigma-Aldrich
Meglumine diatrizoate, organic