Merck
  • Synthesis of Fe3O4 poly(styrene-glycidyl methacrylate) magnetic porous microspheres and application in the immobilization of Klebsiella sp. FD-3 to reduce Fe(III)EDTA in a NO(x) scrubbing solution.
Synthesis of Fe3O4 poly(styrene-glycidyl methacrylate) magnetic porous microspheres and application in the immobilization of Klebsiella sp. FD-3 to reduce Fe(III)EDTA in a NO(x) scrubbing solution.
Bioresource technology (2013-01-22)
Xiaoyan Wang, Zuoming Zhou, Guohua Jing
ABSTRACT
Magnetic poly(styrene-glycidyl methacrylate) porous microspheres (MPPM) with high magnetic contents were prepared by surfactant reverse micelles and emulsion polymerization of monomers, in which the well-dispersed Fe(3)O(4) nanoparticles were modified by polyethylene glycol (PEG) and oleic acid (OA) respectively. The characterizations showed that both of the OA-MPPM and the PEG-MPPM were ferromagnetic, however, the OA-MPPM was used to immobilize the bacteria for more advantages. Therefore, the effects of monomer ratio, surfactant, crosslinker and amount of Fe(3)O(4) on the structure, morphology and magnetic contents of the OA-MPPM were investigated. Then, the OA-MPPM was utilized to immobilize Klebsiella sp. FD-3, an iron-reducing bacterium for Fe(III)EDTA reduction applied in NO(x) removal. Compared with free bacteria, the immobilized FD-3 showed a better tolerance to the unbeneficial pH and temperature conditions.
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)
Supelco
Span® 80, suitable for GC
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, BioReagent, suitable for plant cell culture, crystalline
Sigma-Aldrich
Span® 80, nonionic surfactant