Merck
  • Perturbations of the actin cytoskeleton activate a Dictyostelium STAT signalling pathway.
Perturbations of the actin cytoskeleton activate a Dictyostelium STAT signalling pathway.
European journal of cell biology (2012-03-01)
Tsuyoshi Araki, Jeffrey G Williams
ABSTRACT
The Dictyostelium transcription factor STATc is tyrosine phosphorylated and accumulates in the nucleus when cells are exposed either to hyper-osmotic stress or to the prestalk-inducing polyketide DIF-1. In the case of stress STAT activation is mediated by regulated dephosphorylation; whereby two serine residues on PTP3, the tyrosine phosphatase that de-activates STATc, become phosphorylated after exposure to stress so inhibiting enzymatic activity. We now show that the more highly regulated of the two PTP3 serine residues, S747, is also phosphorylated in response to DIF-1, suggesting a common activation mechanism. Hyper-osmotic stress causes a re-distribution of F-actin to the cortex, cell rounding and shrinkage and we show that DIF-1 induces a similar but transient F-actin re-distribution and rounding response. We also find that two mechanistically distinct inhibitors of actin polymerization, latrunculin A and cytochalasin A induce phosphorylation at S747 of PTP3 and activate STATc. We suggest that PTP3 phosphorylation, and consequent STATc activation, are regulated by changes in F-actin polymerization status during stress and DIF-induced cytoskeletal remodelling.
Sigma-Aldrich
DIF-1, ≥98% (HPLC)